EMAIL THIS PAGE TO A FRIEND
88350 Sigma-Aldrich

Thiamphenicol

≥99.0% (TLC)

Synonym: D-threo-2,2-Dichloro-N-(β-hydroxy-α-[hydroxymethyl]-4-[methylsulfonyl]phenethyl)acetamide

  • CAS Number 15318-45-3

  • Empirical Formula (Hill Notation) C12H15Cl2NO5S

  • Molecular Weight 356.22

  •  Beilstein Registry Number 2819542

  •  EC Number 239-355-3

  •  MDL number MFCD00467983

Purchase

Properties

InChI Key   OTVAEFIXJLOWRX-NXEZZACHSA-N
assay   ≥99.0% (TLC)
mp   163-166 °C
solubility   ethanol: 50 mg/mL, clear, colorless
Mode of action   protein synthesis | interferes

Description

General description

Chemical structure: phenicole

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AB6680000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T0261 Thiamphenicol

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?