EMAIL THIS PAGE TO A FRIEND
89110 Sigma-Aldrich

Thioxanthone

purum, ≥97.0% (HPLC)

Synonym: 9-Oxothioxanthene, Thioxanthen-9-one

  • CAS Number 492-22-8

  • Empirical Formula (Hill Notation) C13H8OS

  • Molecular Weight 212.27

  •  Beilstein Registry Number 140978

  •  EC Number 207-749-4

  •  MDL number MFCD00005066

  •  PubChem Substance ID 329769699

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Other Heterocyclic Building Blocks, S-Containing More...
InChI Key   YRHRIQCWCFGUEQ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (HPLC)
bp   273 °C/715 mmHg(lit.)
mp   207-212 °C
Gene Information   rat ... Maoa(29253), Maob(25750)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?