EMAIL THIS PAGE TO A FRIEND
90794 Sigma-Aldrich

Tributylchlorosilane

Selectophore

Synonym: Chlorotributylsilane, Tributylsilyl chloride

  • CAS Number 995-45-9

  • Linear Formula [CH3(CH2)3]3SiCl

  • Molecular Weight 234.88

  •  Beilstein Registry Number 1742537

  •  EC Number 213-627-1

  •  MDL number MFCD00000509

  •  PubChem Substance ID 329769950

Purchase

Properties

Related Categories Analytical/Chromatography, Chemical Synthesis, Ion Sensor Materials, Protecting and Derivatizing Reagents, Protection and Derivatization,
grade   Selectophore
InChI Key   JSQJUDVTRRCSRU-UHFFFAOYSA-N
refractive index   n20/D 1.446(lit.)
bp   250-252 °C(lit.)
density   0.879 g/mL at 25 °C(lit.)

Description

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2987 8 / PGII
WGK Germany 
1
Flash Point(F) 
206.6 °F
Flash Point(C) 
97 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?