EMAIL THIS PAGE TO A FRIEND
90965 Supelco

sec-Butylamine

analytical standard

Synonym: (±)-2-Aminobutane, (±)-sec-Butylamine

  • CAS Number 13952-84-6

  • Linear Formula CH3CH2CH(NH2)CH3

  • Molecular Weight 73.14

  •  Beilstein/REAXYS Number 1361345

  •  EC Number 237-732-7

  •  MDL number MFCD00008094

  •  PubChem Substance ID 329769975

  •  NACRES NA.24

Purchase

Properties

Related Categories A-B, A-F, All Alphabetically Sorted, Alpha Index of All Pesticide Standards, Analytical Standards,
Quality Level   100
grade   analytical standard
assay   ≥99.0% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤0.5% water
refractive index   n20/D 1.392-1.394
  n20/D 1.3928 (lit.)
bp   63 °C (lit.)
mp   −72 °C (lit.)
density   0.724 g/mL at 25 °C (lit.)
Featured Industry   Agriculture
Cleaning Products
Cosmetics
Environmental
Flavors and Fragrances
Food and Beverages
Personal Care
format   neat
SMILES string   CCC(C)N
InChI   1S/C4H11N/c1-3-4(2)5/h4H,3,5H2,1-2H3
InChI key   BHRZNVHARXXAHW-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN3286 - class 3 - PG 2 - RQ - EHS - basic - Flammable liquid, toxic, corrosive, n.o.s., Hi:ALL(not BR)
WGK Germany 
WGK 2
RTECS 
EO3325000
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Chromatography, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?