• USA Home
  • 92140 - 3,4,5-Trimethoxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND
92140 Sigma-Aldrich

3,4,5-Trimethoxybenzaldehyde

purum, ≥98.0% (HPLC)

  • CAS Number 86-81-7

  • Linear Formula (CH3O)3C6H2CHO

  • Molecular Weight 196.20

  •  Beilstein/REAXYS Number 395163

  •  EC Number 201-701-6

  •  MDL number MFCD00003364

  •  PubChem Substance ID 329769871

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C10-C12, Carbonyl Compounds, Chemical Synthesis,
grade   purum
assay   ≥98.0% (HPLC)
bp   163-165 °C/10 mmHg (lit.)
mp   72-74 °C (lit.)
  73-76 °C
solubility   methanol: 0.1 g/mL, clear
SMILES string   [H]C(=O)c1cc(OC)c(OC)c(OC)c1
InChI   1S/C10H12O4/c1-12-8-4-7(6-11)5-9(13-2)10(8)14-3/h4-6H,1-3H3
InChI key   OPHQOIGEOHXOGX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CU8462000

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

T68403 3,4,5-Trimethoxybenzaldehyde, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?