• USA Home
  • 92302 - Listeria mono Confirmatory Agar (Base)

EMAIL THIS PAGE TO A FRIEND
92302 Millipore

Listeria mono Confirmatory Agar (Base)

for microbiology

Synonym: Listeria monocytogenes Confirmatory Agar (Base)

Purchase

Properties

Related Categories A - M, All Dairy Media (alphabetical), All Food Control Media (alphabetical), All Solid Media/Agars, Listeria,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 12.0 g/L
  B.C. indicator, 8.6 g/L
  disodium hydrogen phosphate anhydrous, 2.5 g/L
  lithium chloride, 10.0 g/L
  α-methyl-D-mannoside, 3.0 g/L
  sodium chloride, 5.0 g/L
  special peptone, 30.0 g/L
  yeast extract, 6.0 g/L
final pH   7.2±0.2 (25 °C)
Featured Industry   Agriculture
Environmental
Food and Beverages
suitability   selective and differential for Listeria spp.

Description

Application

Listeria mono Confirmatory Agar is recommended for the selective and differential isolation of Listeria monocytogenes from clinical and food specimens.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the natural environment, microbes have adapted to the habitats most suitable ...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?