• USA Home
  • 93746 - 2-Methyl-2-[8-nitro-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-4-yl]propan-1-ol

EMAIL THIS PAGE TO A FRIEND
93746 Supelco

2-Methyl-2-[8-nitro-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-4-yl]propan-1-ol

analytical standard

Synonym: 3a,4,5,9b-Tetrahydro-β,β-dimethyl-8-nitro-3H-cyclopenta[c]quinoline-4-ethanol

Purchase

Properties

Related Categories All Doping Standards (A-Z), Anabolic Agent / SARMS / Other, Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol,
grade   analytical standard
InChI Key   CJRZFRLQNHKDCY-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
composition   carbon, 65.3-67.9%
  nitrogen, 9.5-9.9%
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Pharmaceutical (small molecule)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?