• USA Home
  • 94897 - Vinblastine sulfate salt

EMAIL THIS PAGE TO A FRIEND
94897 Sigma-Aldrich

Vinblastine sulfate salt

crystallized, ≥96.0% (TLC)

Synonym: VLB, Vincaleukoblastine sulfate salt

  • CAS Number 143-67-9

  • Empirical Formula (Hill Notation) C46H58N4O9 · H2SO4

  • Molecular Weight 909.05

  •  Beilstein Registry Number 3659812

  •  EC Number 205-606-0

  •  MDL number MFCD00082457

Purchase

Properties

InChI Key   KDQAABAKXDWYSZ-PNYVAJAMSA-N
assay   ≥96.0% (TLC)
quality   crystallized
impurities   ≤15% solvent (ethanol)
mp   267 °C (dec.)(lit.)
storage temp.   2-8°C

Description

Other Notes

Induces microtubule aggregation ; Studies on Tau-tubulin-vinblastine aggregation ; Inhibits monoamine oxidase B.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YY8400000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

V1377 Vinblastine sulfate salt, ≥97% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?