EMAIL THIS PAGE TO A FRIEND
A9306 Sigma-Aldrich

Bovine Serum Albumin

lyophilized powder, low endotoxin, ≥95% (agarose gel electrophoresis)

Synonym: Albumin bovine serum, BSA, Bovine albumin

Purchase

Properties

Related Categories Albumin, Biochemicals and Reagents, Bovine Serum Albumins, Chromatographically Purified, New Zealand Origin Bovine Serum Albumins,
biological source   bovine
assay   ≥95% (agarose gel electrophoresis)
form   lyophilized powder
mol wt   ~66 kDa
purified by   chromatography
origin   New Zealand origin
application(s)   cell culture | mammalian: suitable
impurities   ≤1 EU/mg endotoxin
pH   7
solubility   water: soluble (40 mg/ml)
storage temp.   2-8°C
Gene Information   bovine ... ALB(280717)

Description

Biochem/physiol Actions

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Preparation Note

Purified by chromatography

Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?