• USA Home
  • BCR049 - Benzo[j]fluoranthene

EMAIL THIS PAGE TO A FRIEND
BCR049 Sigma-Aldrich

Benzo[j]fluoranthene

BCR® certified Reference Material

Synonym: Benzo[j]fluoranthene (purity)

  • CAS Number 205-82-3

  • Empirical Formula (Hill Notation) C20H12

  • Molecular Weight 252.31

  •  MDL number MFCD00152264

  •  PubChem Substance ID 329774202

Purchase

Description

Analysis Note

For more information please see:
BCR049

Legal Information

BCR is a registered trademark of European Commission

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles

Articles

EN Method 2005/108/EC: GC Analysis of Pesticide/PAH Compounds on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents EN Method 2005/108/EC: GC Analysis of Pesticide/PAH Compounds on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Pesticides, Purification

GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59
Keywords: Chromatography, Gas chromatography, Mass selective detector

US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608

From our library of Articles, Sigma-Aldrich presents US EPA Method 8100: GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) on SPB®-608
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?