EMAIL THIS PAGE TO A FRIEND
C6520 Sigma-Aldrich

Cannabinol solution

1.0 mg/mL in methanol, analytical standard, for drug analysis

Purchase

Properties

Related Categories β-Glucuronidase Drug Analysis Bundle, Additional Standards, Analytical Standards, Analytical and Industrial Enzymes, Analytical/Chromatography,
grade   analytical standard, for drug analysis
InChI Key   VBGLYOIFKLUMQG-UHFFFAOYSA-N
assay   ≥98%
drug control   Home Office Schedule 1
concentration   1.0 mg/mL in methanol
  1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C
Gene Information   human ... CNR2(1269)
rat ... Cnr1(25248)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Expore Enzymes, Proteins & Cofactors
Protocols & Articles

Articles

HPLC Analysis of Active Cannabinoid Compounds in Gummy Bear Candy on Ascentis® Express Biphenyl with UV Detection after Extraction

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Active Cannabinoid Compounds in Gummy Bear Candy on Ascentis® Express Biphenyl with UV Detection after Extraction
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Cannabinoids on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Cannabinoids on Ascentis® C18
Keywords: Analgesics, Chromatography, High performance liquid chromatography

HPLC Analysis of Cannabinoids using Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Cannabinoids using Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Separation, Solvents

LC/MS Analysis of Cannabinoids on Ascentis® RP-Amide

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Cannabinoids on Ascentis® RP-Amide
Keywords: Analgesics, Chromatography, High performance liquid chromatography, Mass spectrometry, Pharmaceutical

UHPLC Analysis of Cannabinoids on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents UHPLC Analysis of Cannabinoids on Ascentis® Express C18
Keywords: Chromatography, Clinical, Diseases, Forensic, Gas chromatography, High performance liquid chromatography, Mass spectrometry, Pharmaceutical, Separation

UHPLC/MS Analysis of Cannabinoids on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents UHPLC/MS Analysis of Cannabinoids on Ascentis® Express C18
Keywords: Clinical, Diseases, Forensic, Gas chromatography, High performance liquid chromatography, Mass spectrometry, Pharmaceutical, Separation, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?