• USA Home
  • M3282 - Mebeverine hydrochloride

EMAIL THIS PAGE TO A FRIEND
M3282 Sigma-Aldrich

Mebeverine hydrochloride

analytical standard

Purchase

Properties

Related Categories Additional Drugs, Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol
grade   analytical standard
InChI Key   PLGQWYOULXPJRE-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Packaging

10 g in poly bottle

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YX5425000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?