EMAIL THIS PAGE TO A FRIEND
S1503 Sigma-Aldrich

Secobarbital

analytical standard

Synonym: 5-(1-Methylbutyl)-5-(2-propenyl)-2,4,6(1H,3H,5H)-pyrimidinetrione, 5-Allyl-5-(1-methylbutyl)barbituric acid, Quinalbarbitone, Seconal

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Barbiturate Standards, Pharmaceuticals, Illicit Drugs & Alcohol
grade   analytical standard
InChI Key   KQPKPCNLIDLUMF-UHFFFAOYSA-N
form   neat
drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
CP9450000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

HPLC Analysis of Barbiturates in Serum on Discovery® C18 after SPE using Discovery® DSC-18Lt

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates in Serum on Discovery® C18 after SPE using Discovery® DSC-18Lt
Keywords: Calorimetry, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical, Sample preparations, Separation, Solid phase extractions, Solvents

HPLC Analysis of Barbiturates on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Ascentis® C18
Keywords: Anticonvulsants, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical, Separation, Solvents

HPLC Analysis of Barbiturates on Ascentis® C8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Ascentis® C8
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® C18, Unbuffered Mobile Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® C18, Unbuffered Mobile Phase
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® C8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® C8
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

HPLC Analysis of Barbiturates on Discovery® RP-AmideC16

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Barbiturates on Discovery® RP-AmideC16
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?