EMAIL THIS PAGE TO A FRIEND
11600 Sigma

Azocarmine G

for microscopy (Hist.)

Synonym: Acid Red 101, Rosinduline

  • CAS Number 25641-18-3

  • Empirical Formula (Hill Notation) C28H18N3NaO6S2

  • Molecular Weight 579.58

  •  Colour Index Number 50085

  •  EC Number 247-157-3

  •  MDL number MFCD00042002

  •  PubChem Substance ID 57646936

Purchase

Properties

Related Categories A, Cell Biology, Hematology and Histology, Stains & Dyes, A to Z, Stains and Dyes More...
grade   for microscopy (Hist.)
InChI Key   LUERODMRBLNCFK-UHFFFAOYSA-M
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions

Biomedical Applications
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?