EMAIL THIS PAGE TO A FRIEND
52201-013 Sigma-Aldrich

RNAstable® LD

Green Alternative

10mL bottle, for room temp preservation of RNA

Purchase

Properties

Related Categories 12 Principles Aligned Products, Biochemicals and Reagents, Biostabilization Agents, Chemical Synthesis, Greener Alternative Products,
grade   for molecular biology
form   liquid
usage   500 reactions
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
storage temp.   room temp

Description

General description

RNAstable LD allows you to preserve and ship your total RNA, mRNA, and miRNA for up to 12 years at ambient temperatures. They provide a cost-effective, eco-friendly solution to freezer crawl and lab space management. Moreover, they elimnate sample loss due to freeze/thaw cycles. When you are ready to use your RNA samples, you can fully recover them in minutes without any additional purification steps.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product was Designed for Energy Efficiency. To see how Stanford incorporated Biomatrica Technology and saved substantial amounts of energy, see their published study:

Biomatrica technology enables safe storage of biological material at room temperature.

Packaging

1 bottle

Physical form

(1) 10 mL bottle of liquid RNAstable for dry storage.

Legal Information

RNAstable is a registered trademark of Biomatrica

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Biomatrica Selection Guide

Biomatrica products enable researchers in academia, pharmaceutical drug development, and molecular diagnostics to stabilize ship and store biological samples for long periods of time with complete an...
Keywords: Degradations, Diagnostic, Pharmaceutical

RNA Stabilization at Room Temperature in RNAstable®

RNAstable is a novel preservation product developed to protect RNA from degradation during storage or shipment at ambient temperatures. The synthetic storage medium is based on the natural principles...
Keywords: Amplification, Degradations, Gene expression, Microarray Analysis, Polymerase chain reaction, Purification, Sample preparations, Transcription

RNAstable® with Microarray

RNA integrity is a critical factor in obtaining meaningful gene expression data. Current methodologies rely on maintaining samples in cold environments during collection, transport, processing, and s...
Gilberto E. Hernandez, Tony S. Mondala, and Steven R. Head
DNA Array Core Facility, The Scripps Research Institute, La Jolla, California, USA
Keywords: Clinical, Degradations, Diseases, Gene expression, Infectious Diseases, Microarray Analysis, Molecular biology, Phase transitions, Polymerase chain reaction, Precipitation, Purification, RNA purification

Stabilizing RNA at room temperature in RNAstable®

RNAstable® is a novel preservation product developed to protect RNA from degradation during storage or shipment at ambient temperatures. The synthetic storage medium is based on the natural principle...
Sharron Ohgi1, Laurent Coulon, Rolf Muller, Judy-Muller-Cohn, and Omoshile Clement
Biomatrica, Inc., 5627 Oberlin Dr, #120, San Diego, CA 92121, USA.
Keywords: Amplification, Degradations, Gene expression, Hydration reaction, Polymerase chain reaction, Purification, Transcription

miRNA Sequencing with RNAstable® and SeqMatic miRNA kit

From our library of Articles, Sigma-Aldrich presents miRNA Sequencing with RNAstable® and SeqMatic miRNA kit
Keywords: Electrophoresis, Gel electrophoresis, Gene expression, Polymerase chain reaction, Sample preparations, Separation, Sequencing

microRNA (miRNA) Stabilization in RNAstable™

miRNAs play an important role in gene regulation. Genes encoding miRNAs are transcribed and then processed into mature single-stranded stem-loop structures that then bind to complementary mRNA transc...
Keywords: Amplification, Cancer, Cell biology, Degradations, Gene expression, Polymerase chain reaction, Polymerase chain reaction - quantitative, Purification, Sample preparations, Transcription

Related Content

Greener Alternatives Four Categories

A new icon has been added to identify our Greener alternative products. Products with this icon fulfill one of the four criteria.
Keywords: Alternative energy, Environmental, Green chemistry, PAGE, Solvents, Substitutions

Related Products

related product

Product #

Description

Add to Cart

93221-001 RNAstable®, Tube Kit, for room temp preservation of RNA

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?