EMAIL THIS PAGE TO A FRIEND
66777 Sigma

L-Altrose

≥97.0% (HPLC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrate Chemistry, Carbohydrate Derivatives, Carbohydrates, Carbohydrates A to Z,
InChI Key   GZCGUPFRVQAUEE-AZGQCCRYSA-N
assay   ≥97.0% (HPLC)
optical activity   [α]/D -33.0±2.0°, c = 1% in H2O

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

L-Altrose is used as a substrate to identify, differentiate and characterize aldose C-2 epimerases such as cellobiose 2-epimerase from Caldicellulosiruptor saccharolyticus.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?