• USA Home
  • 85580C - LONG® R3 IGF-I human

EMAIL THIS PAGE TO A FRIEND
85580C SAFC

LONG® R3 IGF-I human

media grade, recombinant, expressed in E. coli, lyophilized powder

  • CAS Number 143045-27-6

  • Empirical Formula (Hill Notation) C400H619N111O115S9

  • Molecular Weight 9111.45

  •  NACRES NA.77

Purchase

Properties

Related Categories CHO Platform, Cell Culture, Serum-free Media, Supplements
recombinant   expressed in E. coli
grade   media grade
form   lyophilized powder
availability   not available in Japan
storage temp.   2-8°C

Description

Application

LONG® R3 IGF-I human is suitable in a study to stimulate five Chinese hamster ovary (CHO) clones and explore its effect on the growth, metabolism, and productivity of these clones. It is also suitable for treating murine C2C12 skeletal myoblasts at the time of shifting subconfluent cultures to differentiation medium in a study.

LONG® R3 IGF-I is a recombinant analog of human insulin-like growth factor-I (IGF-I) that has been specifically engineered for the enhancement of cell culture performance. It is more biologically potent in vitro than either insulin or native IGF-I and has been shown to significantly increase recombinant protein production. It is ideal for both research and large-scale culture systems utilizing serum-free or low-level serum applications. LONG R3 IGF-I has been used with numerous cell types including CHO, vaccine-producing cell lines, fibroblasts and stem cells. All cell types that have a growth response to insulin in cell culture have the potential to respond to LONG R3 IGF-I. LONG R3 IGF-I provides equivalent or better performance to recombinant insulin depending on the cell line and clone.

LONG R3 IGF-I is produced in an Escherichia coli expression system without the use of animal-derived components.

Linkage

For more information about LONG®R3 IGF-I, please visit the LONG®R3 IGF-I web page

Other Notes

Please contact your local SAFC sales representative to order.

Legal Information

Available in USA, Canada, and Europe. Please check with your local supplier regarding availability and restrictions in other countries.

LONG is a registered trademark of Repligen Sweden AB

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Pipette Controller from Brand
Protocols & Articles

Articles

LONG®R3IGF-I - GMP Growth Factor Additive for Cell Culture Manufacturing

It is often hard to maintain or improve productivity in serum or animal-free formulations. Many biopharmaceutical manufacturers are trying to eliminate animal-containing raw materials, or move toward...

Keywords: Apoptosis, Cell culture, Cell proliferation, Clinical, Culture media, Diabetes, Fermentation, Gene expression, Growth factors, Infrared spectroscopy, Substitutions, Transduction

LONG®R3IGF-I: An Insulin Alternative Exclusively for Cell Culture Applications

LONG®R3IGF-I is a recombinant analog of human insulin-like growth factor-I (IGF-I) that has been specifically engineered for the enhancement of cell culture performance. LONG®R3IGF-I is more biologic...
Jennifer Fries
BioFiles 2009, 4.5, 14.
Keywords: Aerobic, Apoptosis, Cell culture, Cell proliferation, Clinical, Culture media, Diabetes, Fermentation, Growth factors, High performance liquid chromatography, Infrared spectroscopy, PAGE, Substitutions, Transduction

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

LONG®R3IGF-I

Improving productivity, reducing complexity and mitigating risk is the goal of every Process Development Scientist. For over 20 years, IGF-I has been a proven cGMP cell culture growth factor used LON...
Keywords: Cell culture, Cell proliferation, Culture media, Growth factors

Peer-Reviewed Papers
15

References

Related Products

also commonly purchased with this product

Product #

Description

Add to Cart

14340C EX-CELL® 325 PF CHO Serum-Free Medium for CHO Cells, protein free, liquid, sterile-filtered, suitable for cell culture
24340C EX-CELL® 325 PF CHO Serum-Free Medium for CHO Cells, protein free, dry powder, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?