• USA Home
  • 93140 - 2,3,5-Triphenyltetrazolium chloride

EMAIL THIS PAGE TO A FRIEND
93140 Sigma

2,3,5-Triphenyltetrazolium chloride

≥99.0% (AT)

Synonym: TPTZ, TTZ, Tetrazolium Red

  • CAS Number 298-96-4

  • Empirical Formula (Hill Notation) C19H15ClN4

  • Molecular Weight 334.80

  •  Beilstein Registry Number 3923356

  •  EC Number 206-071-6

  •  MDL number MFCD00011963

  •  PubChem Substance ID 329770208

Purchase

Description

Other Notes

Review: Tetrazolium salts and formazans

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XF8100000

Expore Enzymes, Proteins & Cofactors

Biomedical Applications
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?