• USA Home
  • A4430 - Amobarbital sodium salt

EMAIL THIS PAGE TO A FRIEND
A4430 Sigma

Amobarbital sodium salt

Synonym: 5-Ethyl-5-isoamylbarbituric acid, Amytal

Purchase

Properties

InChI Key   URLVKOXOUJHSEB-UHFFFAOYSA-N
drug control   USDEA Schedule II; Home Office Schedule 3; psychotrope; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

Description

Biochem/physiol Actions

Sedative/hypnotic with rapid onset and short duration of action; can be used to terminate acutely generated convulsions.

General description

Amobarbital sodium salt is a di-alkyl substituted oxybarbiturate, which is used as a hypnotic and sedative.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
CQ5250000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?