• USA Home
  • A5525 - 5′-Amino-5′-deoxythymidine

EMAIL THIS PAGE TO A FRIEND
A5525 Sigma-Aldrich

5′-Amino-5′-deoxythymidine

  • CAS Number 25152-20-9

  • Empirical Formula (Hill Notation) C10H15N3O4

  • Molecular Weight 241.24

  •  MDL number MFCD00047488

  •  PubChem Substance ID 24890985

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   PYWLBQPICCQJFF-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Application

5′-Amino-5′-deoxythymidine may be used as a substrate in the synthesis of amide-conjugated nucleosides and their analogues. 5′-Amino-5′-deoxythymidine may be used for DNA 5′-terminal site-specific modification. 5′-Amino-5′-deoxythymidine is used to produce modified thymidine dimer oligonucleotide building units (squaryl groups) to create new oligonucleotide analogues.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?