• USA Home
  • A8352 - Adenosine Transporter Receptor human

EMAIL THIS PAGE TO A FRIEND
A8352 Sigma-Aldrich

Adenosine Transporter Receptor human

membrane suspension

Synonym: Nucleoside Transporter 1

Purchase

Properties

form   membrane suspension
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... SLC29A1(2030)

Description

Biochem/physiol Actions

Membranes suspended in 50 mM Tris-HCl containing10% sucrose, pH 7.4. Actual concentration and specific binding capacity are provided with each lot.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Fluorescent G protein-coupled receptors (GPCRs)

G protein-coupled receptors (GPCRs) are the largest class of transmembrane proteins and the targets for almost half of the clinical drugs in the market today. Advances in X-ray crystallography and ot...
Mike Earley, Market Segment Manager
Biofiles Vol. 8, No. 4
Keywords: Clinical, Confocal microscopy, Ligands, Microscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?