• USA Home
  • A9879 - 2-Amino-2-methyl-1-propanol

EMAIL THIS PAGE TO A FRIEND
A9879 Sigma

2-Amino-2-methyl-1-propanol

≥95%, syrup

Synonym: β-Aminoisobutyl alcohol, AMP 95, AMP

  • CAS Number 124-68-5

  • Linear Formula (CH3)2C(NH2)CH2OH

  • Molecular Weight 89.14

  •  Beilstein Registry Number 505979

  •  EC Number 204-709-8

  •  MDL number MFCD00008051

Purchase

Properties

vapor density   3 (vs air)
vapor pressure   <1 mmHg ( 25 °C)
InChI Key   CBTVGIZVANVGBH-UHFFFAOYSA-N
assay   ≥95%
form   syrup
refractive index   n20/D 1.4455(lit.)
useful pH range   9.0 - 10.5
pKa (25 °C)   9.7
bp   165 °C(lit.)
mp   24-28 °C(lit.)
density   0.934 g/mL at 25 °C(lit.)

Description

Physical form

Syrup at room temperature

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
154.4 °F
Flash Point(C) 
68 °C
Protocols & Articles

Related Content

Buffer Reference Center

Citric Acid – Na2HPO4 Buffer Solutions, pH approx. 2.6–7.61 Citric acid monohydrate, C6H8O7 • H2O, M. wt. 210.14; 0.1M-solution contains 21.01 g/l. Na2HPO4, M. wt. 141.98; 0.2M-solution contains 28.4...
Keywords: Biological Buffers, Buffers

缓冲液参考中心

柠檬酸 – Na2HPO4 缓冲液,pH约为 2.6–7.61 柠檬酸一水合物, C6H8O7 • H2O, M. wt. 210.14; 0.1M的溶液中含有21.01 g/l。 Na2HPO4, M. wt. 141.98; 0.2M的溶液中含有28.40g / l 或 Na2HPO4 • 2H2O, M. wt. 178.05; 0.2M的溶液中含有35.61 g/l。 xml 0.1M的...

Related Products

replaced by

Product #

Description

Add to Cart

A65182 2-Amino-2-methyl-1-propanol, ~5% Water, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?