• USA Home
  • B124 - Bufotenine oxalate salt

EMAIL THIS PAGE TO A FRIEND
B124 Sigma

Bufotenine oxalate salt

  • CAS Number 16698-26-3

  • Empirical Formula (Hill Notation) C12H16N2O · C2H2O4

  • Molecular Weight 294.30

Purchase

Properties

drug control   USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN2 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?