• USA Home
  • C3811 - Centrophenoxine hydrochloride

EMAIL THIS PAGE TO A FRIEND
C3811 Sigma

Centrophenoxine hydrochloride

plant cell culture tested

Synonym: (4-Chlorophenoxy)acetic acid 2-(dimethylamino)ethyl ester hydrochloride, 2-(Dimethylamino)ethyl 4-chlorophenoxyacetate hydrochloride, Meclofenoxate hydrochloride

Purchase

Properties

InChI Key   FIVHOHCAXWQPGC-UHFFFAOYSA-N
suitability   plant cell culture tested
storage temp.   −20°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AG0440000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?