• USA Home
  • C6383 - 5-Chloro-8-quinolinol

EMAIL THIS PAGE TO A FRIEND
C6383 Sigma-Aldrich

5-Chloro-8-quinolinol

Synonym: 5-Chloro-8-hydroxyquinoline

  • CAS Number 130-16-5

  • Empirical Formula (Hill Notation) C9H6ClNO

  • Molecular Weight 179.60

  •  Beilstein/REAXYS Number 5289

  •  EC Number 204-978-1

  •  MDL number MFCD00006788

  •  PubChem Substance ID 329775123

Purchase

Properties

mp   122-124 °C (lit.)
SMILES string   Oc1ccc(Cl)c2cccnc12
InChI   1S/C9H6ClNO/c10-7-3-4-8(12)9-6(7)2-1-5-11-9/h1-5,12H
InChI key   CTQMJYWDVABFRZ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
VC4590000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C47000 5-Chloro-8-quinolinol, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?