• USA Home
  • CKH200A - Tumor Necrosis Factor-α ELISA Kit, Human

EMAIL THIS PAGE TO A FRIEND
CKH200A Sigma

Tumor Necrosis Factor-α ELISA Kit, Human

Purchase

Properties

shipped in   wet ice
storage temp.   2-8°C

Description

Other Notes

The Growth Factor ELISA assay is designed to measure growth factor levels in tissue culture media, serum, plasma and other biological fluids. The ELISA assay uses the "sandwich" enzyme immunoassay technique, with a monoclonal primary capture antibody and a peroxidase labeled specific secondary antibody. The monoclonal primary capture antibody is pre-coated onto a 96-well plate with removable strips. The ELISA kits detect picogram quantities of cytokines. It is a solid phase ELISA which measures TNF-α in less than 4 hours for tissue culture samples and less than 5 hours using serum, plasma or other biological fluids.

Specificity

This ELISA kit has been shown to accurately quantitate recombinant human TNF-α and can be used to determine relative mass values for natural human TNF-α.

Standard component

Description

Product #

Add to Cart

96 well microtiter plate coated with antibody 1 ea    
Safety & Documentation

Safety Information

RIDADR 
UN 1830 8/PG 2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?