• USA Home
  • CS0770 - NDPase (GDPase, UDPase, ADPase) Assay Kit

EMAIL THIS PAGE TO A FRIEND
CS0770 Sigma

NDPase (GDPase, UDPase, ADPase) Assay Kit

sufficient for 300 assays (96-well plates)

Purchase

Properties

Related Categories Cell Analysis, Cell Biology, Cell Signaling and Neuroscience, Cytoskeleton & Extracellular Matrix Kits, Cytoskeleton and Extracellular Matrix More...
usage   sufficient for 300 assays (96-well plates)
shipped in   dry ice
storage temp.   −20°C

Description

Analysis Note

The kit was tested on rat tissue extracts (liver, kidney, spleen, and heart) and on a yeast (S. cerevisae) extract. It was also tested with microsomal fractions prepared from rat tissues and cell lines.

Application

A simple and reliable method for measuring NDPase activity in sub-cellular fractions based on the detection of the inorganic phosphate released from NDP by the appropriate NDPase. The color formation, measured at 700 nm in the presence of molybdate, is proportional to the amount of the inorganic phosphate released by enzymes.

Unit Definition

1 unit of NDPase releases 1 μmole of inorganic phosphate per minute at 25 °C under conditions specified by the assay.

Biochem/physiol Actions

GDPases/UDPases (NDPases) in yeasts are located exclusively in the Golgi complex and are a convenient marker for Golgi detection. NDPs are degraded to NMPs by nucleoside diphosphatase (GDPase/UDPase). The NMPs are then used in antiport transport of new nucleotide sugars into the organelle lumen. ADPases are known to be expressed on the cell plasma membrane (endothelial cells and vascular precursors) where they participate in nucleotide metabolism.

Price and Availability


Flow Cytometry

Milli-Q® Water Purification Solutions

Kit component also available separately

Description

Product #

Add to Cart

Sodium dodecyl sulfate Solution 10% 5 mL SDS    
Phosphorus standard Solution 0.65 mM 1.7 mL SDS    

Kit component only

Description

Product #

Add to Cart

ADPase Assay Reagent 10 mL SDS    
GDPase Assay Reagent 10 mL SDS    
UDPase Assay Reagent 10 mL SDS    
Ascorbic Acid Solution 1 mL SDS    
Molybdenum Color Reagent 60 mL SDS    
Safety & Documentation

Safety Information

RIDADR 
UN 3264 9/PG 3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?