• USA Home
  • CUSTOMCELLS - Custom Cell Lines

EMAIL THIS PAGE TO A FRIEND
CUSTOMCELLS

Custom Cell Lines

Engineered Cell Lines from Cell Design Studio

Synonym: CRISPR, SNP, ZFN, cell design studio, cell line, custom cell line development, custom cell line engineering, diagnostic, disease, engineered custom cell lines, engineered, gene disruption, iPS, knockin, knockout, targeted integration

Choose from the products below and click the Inquire button...need more help? more

  • 1. Decide which product type best fits your needs.
  • 2. Click the "Inquire Here" button to contact a Cell Design Studio Team member about your project..

Gene Knockout Cell Line

  • Cell line provided by customer or utilization of in-house Sigma cell line
  • Evaluation of gene target and target cell line including feasibility and risk assessment
  • Design and assembly of either Zinc Finger Nuclease or CRISPR reagent
  • Validation of nuclease in target cell line
  • Isolation of clonal populations
  • Identification of targeted gene disruption
  • Cryopreservation and cell line delivery
  • Learn more about custom cell lines and optional assays.

Gene Modification Cell Line

  • Cell line provided by customer or utilization of in-house Sigma cell line
  • Evaluation of gene target and target cell line including feasibility and risk assessment
  • Design and assembly of either Zinc Finger Nuclease or CRISPR reagent
  • Design,assembly, and construction of donor plasmid or oligo
  • Validation of nuclease in target cell line
  • Isolation of clonal populations
  • Identification of targeted gene modification
  • Cryopreservation and cell line delivery
  • Learn more about custom cell lines and optional assays.

Targeted Integration Cell Line

  • Cell line provided by customer or utilization of in-house Sigma cell line
  • Evaluation of gene target and target cell line including feasibility and risk assessment
  • Design and assembly of either Zinc Finger Nuclease or CRISPR reagent targeting a safe harbour locus: AAVSI (human), ROSA26 (mouse/rat)
  • Design, assembly, and construction of donor plasmid or oligo
  • Validation of nuclease in target cell line
  • Isolation of clonal populations
  • Identification of targeted integration
  • Cryopreservation and cell line delivery
  • Learn more about custom cell lines and optional assays.

Reporter Cell Line

  • Cell line provided by customer or utilization of in-house Sigma cell line
  • Evaluation of gene target and target cell line including feasibility and risk assessment
  • Design and assembly of either Zinc Finger Nuclease or CRISPR reagent
  • Design,assembly, and construction of donor plasmid
  • Validation of nuclease in target cell line
  • Isolation of clonal populations
  • Identification of targeted integration
  • Cryopreservation and cell line delivery
  • Reporter lines currently available
  • Learn more about custom cell lines and optional assays.

Gene Knockdown Pool

  • Cell line provided by customer or utilization of in-house Sigma cell line
  • Evaluation of gene target and target cell line including feasibility and risk assessment
  • Design and assembly of shRNA viral particles
  • Design,assembly, and construction of donor plasmid or oligo if required
  • Validation of nuclease in target cell line
  • Cryopreservation and pool delivery
  • Learn more about custom cell lines and optional assays.

Pool of modified cells

  • Cell line provided by customer or utilization of in-house Sigma cell line
  • Evaluation of gene target and target cell line including feasibility and risk assessment
  • Design and assembly of either Zinc Finger Nuclease or CRISPR reagent
  • Design, assembly, and construction of donor plasmid or oligo if required
  • Validation of nuclease in target cell line
  • Cryopreservation and pool delivery
  • Learn more about custom cell lines and optional assays.

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?