• USA Home
  • D5008 - 2-Dimethylamino-6-hydroxypurine

EMAIL THIS PAGE TO A FRIEND
D5008 Sigma

2-Dimethylamino-6-hydroxypurine

≥98%

Synonym: N2,N2-Dimethylguanine

  • CAS Number 1445-15-4

  • Empirical Formula (Hill Notation) C7H9N5O

  • Molecular Weight 179.18

  •  MDL number MFCD00055968

  •  PubChem Substance ID 24893914

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   XMSMHKMPBNTBOD-UHFFFAOYSA-N
assay   ≥98%
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?