• USA Home
  • E9533 - pFLAG−Shift12c Expression Vector

EMAIL THIS PAGE TO A FRIEND
E9533 Sigma

pFLAG−Shift12c Expression Vector

Bacterial vector for cytoplasmic expression of N-terminal FLAG-Shift fusion proteins

Purchase

Properties

grade   for molecular biology
form   buffered aqueous solution
conjugate   FLAG® tagged
shipped in   dry ice
storage temp.   −20°C

Description

Application

Cytoplasmic expression of N-terminal FLAG® fusion proteins under control of the tac promoter. The FLAG®-Shift DNA sequence causes expression of FLAG® fusion proteins in all three reading frames and is ideal when the reading frame of the insert is unknown. Supplied with a pFLAG-Shift12c-BAP Control Vector.

Expression vector and control vector sequences can be found here.

Other Notes

Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Sigma-Aldrich Co. LLC

pFLAG-SHIFT is a trademark of Sigma-Aldrich Co. LLC

Kit component only

Description

Product #

Add to Cart

pFLAG−Shift12c Expression Vector SDS    
FLAG-Shift12−BAP Control plasmid SDS    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?