• USA Home
  • G4506 - β-Glucuronidase–Agarose.

EMAIL THIS PAGE TO A FRIEND
G4506 Sigma-Aldrich

β-Glucuronidase–Agarose.

ammonium sulfate suspension, 200,000-400,000 units/g agarose, pH 5.0 (One ml gel yields approx. 10,000 units), enzyme from bovine liver

Synonym: β-D-Glucuronide glucuronosohydrolase

Purchase

Properties

biological source   enzyme from bovine liver
form   ammonium sulfate suspension
specific activity   200,000-400,000 units/g agarose, pH 5.0 (One ml gel yields approx. 10,000 units)
matrix   beaded agarose
storage temp.   2-8°C

Description

Unit Definition

One Sigma or modified "Fishman" unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37°C at the appropriate pH (30 min assay).

Quantity

One modified "Fishman" unit equals approx. 5 × 10−5 old μMolar unit.

Physical form

Suspension in 2.0 M (NH4)2SO4, pH 7.0

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Enzymes & Proteins

Committed to developing and providing breakthrough problem-solving reagents and solutions, explore our comprehensive offering of enzymes and protein reagents and kits with the confidence that you wil...
Keywords: Cell signaling, Diagnostic, Enzyme-linked immunosorbent assay, Immunohistochemistry, Metabolism, Molecular biology, Purification, Western blot

Related Products

substrate

Product #

Description

Add to Cart

N7006 4-Nitrophenyl β-D-glucopyranoside, ≥98% (TLC)
B8174 5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, tablet
B4532 5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?