• USA Home
  • GE17-0885-01 - Gammabind G Sepharose®

EMAIL THIS PAGE TO A FRIEND
GE17-0885-01

Gammabind G Sepharose®

GE Healthcare, 17-0885-01, pack of 5 mL

  •  NACRES NA.56

Purchase

Properties

Related Categories Antibodies, Antibody Purification and Characterization, Protein A, G and L Resins, Supplementary Products
shelf life   Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
packaging   pack of 5 mL
mfr. no.   GE Healthcare, 17-0885-01
matrix   4% agarose
particle size   45-165 μm
average diameter   90 μm
cleaning in place   2 - 9 (pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.)
working range   3 - 9
capacity   >17 mg binding capacity(human IgG/ml)
storage temp.   2-8°C

Description

General description

Recombinant GammaBind is an affinity medium for laboratory scale antibody purification. Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse.

GammaBind G Sepharose® is GammaBind G, Type 2, covalently immobilized to Sepharose® 4B by the cyanogen bromide method. GammaBind G, Type 2, a recombinant form of streptococcal Protein G, binds to the Fc region of IgG from a variety of mammalian species. GammaBind G Sepharose® may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.

GammaBind G Sepharose® may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium. Since only the Fc region is involved in binding, the Fab region is still available for binding antigen. Hence, GammaBind G Sepharose® is extremely useful for isolation of immune complexes. GammaBind G will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin.

Application

The potential applications of GammaBind G Sepharose® include practically all of the current and projected applications of Protein A Sepharose. However, GammaBind G and Protein A have different IgG binding specificities, depending on the origin of the IgG. Compared to Protein A, GammaBind G binds more strongly to mouse and rat monoclonal IgG and to IgG from goat, sheep, horse, cow, human, rabbit and other mammalian species.

Features and Benefits

• Recombinant GammaBind G type 2 coupled to Sepharose® 4B engineered for antibody purification at laboratory scale.
• Binds to all IgG subclasses from human, mouse, and rat.
• Binds total IgG from guinea pig, rabbit, goat, cow sheep, and horse.
• Will not cross-react with other serum proteins such as IgM, IgE, IgA or transferrin.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.gelifesciences.com.

Legal Information

Sepharose is a registered trademark of Cytiva

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?