• USA Home
  • GE28-4055-01 - GST MultiTrap Fast Flow

EMAIL THIS PAGE TO A FRIEND
GE28-4055-01

GST MultiTrap Fast Flow

GE Healthcare, 28-4055-01

Purchase

Properties

Related Categories GE Healthcare Life Sciences Protein Sample Preparation Formats, Molecular Biology, MultiTrap Centrifuge/vacuum, Native Protein Sample Preparation, Proteomics More...
material   polypropylene and polyethylene
shelf life   Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
mfr. no.   GE Healthcare, 28-4055-01
size   127.8 mm × 85.5 mm × 30.6 mm
matrix   highly cross-linked 4% agarose
average diameter   90 μm
cleaning   3 - 12
working range   3 - 12
capacity   >10 mg/mL binding capacity(Binding of GST to glutathione is flow-rate dependent and lower flow rates often increase the binding capacity. This is important to consider during sample loading and elution.)

Description

General description

GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and rapid parallel purification of glutathione S-transferase (GST) tagged proteins.

Application

GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and purification of glutathione S-transferase (GST) tagged proteins.

Purification of up to 0.5 mg of GST-tagged Proteins/well directly from unclarified cell lysate can be achieved using GST MultiTrap filter plates. Direct purification of GST-tagged Proteins from unclarified lysate shortens handling time and minimizes degradation of sensitive target Proteins. The 96-well plate format gives great flexibility, both when working with automated robotic systems and manually using centrifugation or vacuum. Consistent well-to-well and plate-to-plate performance ensures high reproducibility.

GST MultiTrap FF is prepacked with Glutathione Sepharose® 4 Fast Flow, while GST MultiTrap 4B is prepacked with Glutathione Sepharose® 4B.

Features and Benefits

• 96-well filter plate format for high-throughput screening and parallel purification of GST-tagged* Proteins.
• Highly reproducible well-to well and plate-to-plate results.
• High purity in one step, high chemical stability. Load unclarified samples directly to the wells.
• Wells are prepacked with Glutathione Sepharose® media for convenience.
• Easy and predictable scale-up using prepacked GSTrap FF, GSTPrep FF 16/10, and GSTrap 4B columns.

Storage and Stability

Store at 4 to 30 °C (20% Ethanol)

Analysis Note

www.gelifesciences.com.

Legal Information

GSTPrep is a trademark of Cytiva

GSTrap is a trademark of Cytiva

MultiTrap is a trademark of Cytiva

Sepharose is a registered trademark of Cytiva

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Protocols

Characteristics of Glutathione Sepharose Products

Appendix 2, Extracted from Affinity Chromatography Vol. 2: Tagged Proteins, GE Healthcare, 2016  
Keywords: Affinity chromatography, Buffers, Centrifugation, Chromatography, Inductively coupled plasma, Ligands, Purification, Tagged proteins

Characteristics of Glutathione Sepharose and HiTrap Benzamidine FF (High Sub) Media and Columns

Glutathione Sepharose High Performance is recommended for high-resolution purification of GST-tagged proteins, providing sharp peaks and concentrated eluent. Glutathione Sepharose 4 Fast Flow is exce...
Keywords: Buffers, Centrifugation, Chromatography, Inductively coupled plasma, Ligands, Purification, Tagged proteins

Desalting/Buffer Exchange and Concentration for Affinity Chromatography of Tagged Proteins

Desalting at laboratory scale is a well-proven, simple, and very fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired bu...
Keywords: Absorption, Addition reactions, Affinity chromatography, Buffers, Centrifugation, Chromatography, Detergents, Dialysis, Fractionation, Ion Exchange, PAGE, Precipitation, Sample preparations, Separation, Size-exclusion chromatography, Tagged proteins

High-throughput Screening Using GST MultiTrap FF and GST MultiTrap 4B 96-well Filter Plates

GST MultiTrap FF and GST MultiTrap 4B (Fig 3.4) are prepacked, disposable 96-well filter plates for reproducible, high-throughput screening of GST-tagged proteins. The plates are filled with a define...
Keywords: Buffers, Cell disruption, Centrifugation, Filtration, Gene expression, Oxidations, PAGE, Purification, Sample preparations, Sonication, Tagged proteins

Manual and Automated Purification

Chapter 2, Extracted from Affinity Chromatography Vol. 2: Tagged Proteins, GE Healthcare, 2016
Keywords: Affinity chromatography, Centrifugation, Chromatography, Gene expression, Tagged proteins

Preparative Purification Using GSTPrep FF 16/10 Column

GSTPrep FF 16/10 columns are based on the 20 ml HiPrep column design, ready to use for easy, one-step preparative purification of GST-tagged proteins, other glutathione S-transferases, and glutathion...
Keywords: Addition reactions, Buffers, Centrifugation, Chromatography, Homogenization, Oxidations, PAGE, Purification, Sample preparations, Sonication, Tagged proteins

Removal of GST tag by enzymatic cleavage

Removal of the GST tag is often necessary to be able to perform functional or structural studies of the target protein. Tagged proteins containing a PreScission Protease, thrombin, or Factor Xa recog...
Keywords: Absorption, Affinity chromatography, Buffers, Cell disruption, Centrifugation, Chromatography, Digestions, PAGE, Size-exclusion chromatography, Sonication, Tagged proteins

Starting materials needed for the purification of integral membrane proteins for structural and functional studies

Extracted from Purifying Challenging Proteins - Principles and Methods, GE Healthcare, 2007
Keywords: Cloning, Detergents, Gene expression, Glycosylations, PAGE, Positron Emission Tomography, Separation, Tagged proteins

Troubleshooting Strategies in Purification of GST-tagged Proteins

This troubleshooting guide addresses the common problems associated with the majority of purification methods using the different Glutathione Sepharose media.
Keywords: Buffers, Cell disruption, Centrifugation, Degradations, Detergents, Electrophoresis, Gene expression, Ion Exchange, Precipitation, Purification, Sonication, Tagged proteins, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?