EMAIL THIS PAGE TO A FRIEND
H3137 Sigma

HEPPSO Hydrate

≥99% (titration)

Synonym: 4-(2-Hydroxyethyl)piperazine-1-(2-hydroxypropanesulfonic acid) Hydrate hydrate

  • Linear Formula C9H20N2O5S.xH2O

  • Molecular Weight 268.33 (anhydrous basis)

  •  Beilstein Registry Number 798996

  •  MDL number MFCD00038351

  •  PubChem Substance ID 24895555

Purchase

Properties

assay   ≥99% (titration)
useful pH range   7.1 - 8.5
pKa (25 °C)   7.5
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Buffer Reference Center

Citric Acid – Na2HPO4 Buffer Solutions, pH approx. 2.6–7.61 Citric acid monohydrate, C6H8O7 • H2O, M. wt. 210.14; 0.1M-solution contains 21.01 g/l. Na2HPO4, M. wt. 141.98; 0.2M-solution contains 28.4...
Keywords: Biological Buffers, Buffers

缓冲液参考中心

柠檬酸 – Na2HPO4 缓冲液,pH约为 2.6–7.61 柠檬酸一水合物, C6H8O7 • H2O, M. wt. 210.14; 0.1M的溶液中含有21.01 g/l。 Na2HPO4, M. wt. 141.98; 0.2M的溶液中含有28.40g / l 或 Na2HPO4 • 2H2O, M. wt. 178.05; 0.2M的溶液中含有35.61 g/l。 xml 0.1M的...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?