• USA Home
  • H4643 - (S)-(−)-3-(3-Hydroxyphenyl)-N-propylpiperidine hydrochloride

EMAIL THIS PAGE TO A FRIEND
H4643 Sigma-Aldrich

(S)-(−)-3-(3-Hydroxyphenyl)-N-propylpiperidine hydrochloride

Synonym: Preclamol, S-(−)-3-PPP

  • CAS Number 88768-67-6

  • Empirical Formula (Hill Notation) C14H21NO · HCl

  • Molecular Weight 255.78

  •  MDL number MFCD00069366

  •  PubChem Substance ID 329815128

Purchase

Properties

InChI Key   NRHUDETYKUBQJT-BTQNPOSSSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?