• USA Home
  • I2510 - Iscove’s Modified Dulbecco’s Medium

EMAIL THIS PAGE TO A FRIEND
I2510 Sigma-Aldrich

Iscove’s Modified Dulbecco’s Medium

suitable for hybridoma, powder, Hybri-Max

Synonym: IMDM

Purchase

Properties

form   powder
application(s)   cell culture | hybridoma: suitable
impurities   endotoxin, tested
storage temp.   2-8°C

Description

Application

Reported to support murine B lymphocytes, hematopoietic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes, and a variety of hybrid cells.

Formula variant

With L-glutamine and 25 mM HEPES. Without sodium bicarbonate.

Quantity

Formulated to contain 17.7 grams of powder per liter of medium.

Reconstitution

Supplement with 3.024 g/L sodium bicarbonate.

Legal Information

Hybri-Max is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

I7633 Iscove’s Modified Dulbecco’s Medium, With L-glutamine and 25 mM HEPES, without sodium bicarbonate, powder, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?