• USA Home
  • I5250 - myo-Inositol 2-monophosphate bis(cyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
I5250 Sigma-Aldrich

myo-Inositol 2-monophosphate bis(cyclohexylammonium) salt

~95%, powder

  • CAS Number 103529-92-6

  • Empirical Formula (Hill Notation) C6H13O9P · 2C6H13N

  • Molecular Weight 458.48

  •  Beilstein/REAXYS Number 3923086

  •  MDL number MFCD00055895

  •  PubChem Substance ID 329815382

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates H-L, Monosaccharide More...
assay   ~95%
form   powder
color   white
SMILES string   NC1CCCCC1.NC2CCCCC2.O[C@@H]3[C@@H](O)[C@H](O)[C@H](OP(O)(O)=O)[C@H](O)[C@H]3O
InChI   1S/2C6H13N.C6H13O9P/c2*7-6-4-2-1-3-5-6;7-1-2(8)4(10)6(5(11)3(1)9)15-16(12,13)14/h2*6H,1-5,7H2;1-11H,(H2,12,13,14)/t;;1-,2-,3+,4+,5-,6+
InChI key   IMIHZMWNWGIJKQ-YAUWMCRJSA-N

Description

Packaging

10, 50 mg in poly bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?