• USA Home
  • L4131 - Lanthanum(III) chloride heptahydrate

EMAIL THIS PAGE TO A FRIEND
L4131 Sigma

Lanthanum(III) chloride heptahydrate

≥99.9%

Purchase

Properties

InChI Key   FDFPDGIMPRFRJP-UHFFFAOYSA-K
assay   ≥99.9%
mp   91 °C (dec.) (lit.)

Description

Biochem/physiol Actions

Calcium channel blocker

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?