• USA Home
  • L8792 - Lofepramine hydrochloride

EMAIL THIS PAGE TO A FRIEND
L8792 Sigma

Lofepramine hydrochloride

≥98% (HPLC), solid

Synonym: N-(4-Chlorobenzoylmethyl)-3-(10,11-dihydro-5h-dibenz[b,f]azepin-5-yl)-n-methylpropylamine hydrochloride

Purchase

Description

Biochem/physiol Actions

Lofepramine is an antidepressant; serotonin and norepinephrine re-uptake inhibitor (SNRI).

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AM6485000

Documents

Certificate of Analysis


Discover your Target

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Serotonin Receptors

Serotonin (5-hydroxytryptamine, 5-HT) is widely distributed throughout the mammalian body, being synthesized from L-tryptophan in enterochromaffin cells of the gastro-intestinal tract as well as in s...
Keywords: Active transport, Atomic absorption spectroscopy, Cancer, Clinical, Gene expression, Ligands, Neurotransmitters, Obesity, Polymorphisms, Schizophrenia, Transduction

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?