• USA Home
  • M0156 - Corning® 96 Well EIA/RIA Assay Microplate
EMAIL THIS PAGE TO A FRIEND
M0156 Sigma
Corning® 96 Well EIA/RIA Assay Microplate
flat bottom (medium-binding)
Purchase
Properties
material   flat bottom (medium-binding)
  polystyrene
mfr. no.   Corning 9017
size   96 wells , 96 well plate
well volume   330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL
well working volume   75-200 μL
suitability   suitable for (EIA/RIA; immunoassays)
Description
General description
These 96 well EIA/RIA plates are designed for in vitro diagnostic immunoassays. All EIA/RIA plates are manufactured from special medical-grade polystyrene for uniform binding, high optical clarity and low background absorption. In addition, the Easy-Wash products are part of the Certified Surface Chemistry Programme, which guarantees protein binding performance to be within QA release specifications.
Legal Information
Corning is a registered trademark of Corning, Inc.
Easy-Wash is a trademark of Corning, Inc.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Content
Cell Culture Troubleshooting Guide
Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions
Related Products
replaced by
Product #
Description
Add to Cart
CLS9017 Corning® 96 Well EIA/RIA Assay Microplate, 96 well plate, non-treated (medium bind), polystyrene, flat bottom, clear, non-sterile, 100/cs
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?