EMAIL THIS PAGE TO A FRIEND
M0521 Sigma

Medazepam

Synonym: 7-Chloro-2,3-dihydro-1-methyl-5-phenyl-1H-1,4-benzodiazepine

Purchase

Properties

drug control   USDEA Schedule IV;
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
DE9070000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?