EMAIL THIS PAGE TO A FRIEND
M3274 SAFC

Medium 199

powder, Modified, suitable for cell culture

Purchase

Properties

form   powder
IVD   for in vitro diagnostic use
application(s)   cell culture | mammalian: suitable
storage temp.   2-8°C

Description

Application

Originally developed as a completely defined media formulation for the culture of primary explants. These media have broad applicability, particularly for non-transformed cells and are widely used for vaccine production, primary pancreatic explants, and lens tissues.

Formula variant

With Hanks′ salts. Without L-glutamine, phenol red and sodium bicarbonate.

Quantity

Formulated to contain 10.9 grams of powder per liter of medium.

Reconstitution

Supplement with 0.1 g/L L-glutamine, 0.35 g/L sodium bicarbonate.

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?