EMAIL THIS PAGE TO A FRIEND
M8510 Sigma

1-Methylhypoxanthine

~98%

Synonym: 6-Hydroxy-1-methylpurine

Purchase

Properties

assay   ~98%
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?