EMAIL THIS PAGE TO A FRIEND
O2627 Sigma-Aldrich

Ouabagenin

≥95%

Synonym: 1β,3β,5,11α,14,19-Hexahydroxy-5β-card-20(22)-enolide, G-Strophanthidin

  • CAS Number 508-52-1

  • Empirical Formula (Hill Notation) C23H34O8

  • Molecular Weight 438.51

  •  MDL number MFCD00057697

  •  PubChem Substance ID 24897964

Purchase

Properties

InChI Key   BXSABLKMKAINIU-QOHCMMFCSA-N
assay   ≥95%
storage temp.   2-8°C

Description

Biochem/physiol Actions

Ouabagenin exerts cardiotonic effects through inhibition of Na+/K+-ATPase like the glycosides, albeit at significantly lower potency.

General description

The aglycone of the cardiac glycoside ouabain.

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. O2627.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGI
WGK Germany 
3
RTECS 
FH5100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?