EMAIL THIS PAGE TO A FRIEND
O9640 Sigma

Oleandrin

≥98% (HPLC)

Synonym: 5β,20(22)-Cardenolide-3β,14,16β-triol-3-([2,6-dideoxy-3-O-methyl-α-L-arabinohexopyranosyl]oxy) 16-acetate

  • CAS Number 465-16-7

  • Empirical Formula (Hill Notation) C32H48O9

  • Molecular Weight 576.72

  •  EC Number 207-361-5

  •  PubChem Substance ID 329818943

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Ion Channels, Ion Pump Inhibitors, Monovalent Ion Channels,
InChI Key   JLPDBLFIVFSOCC-XYXFTTADSA-N
assay   ≥98% (HPLC)
form   powder
color   white to off-white
solubility   DMSO: >20 mg/mL
storage temp.   room temp

Description

Biochem/physiol Actions

Cardiac glycoside, inhibits Na(+)/K(+) ATPase; potent carcinostatic cardiac glycoside.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
FH4585000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Milli-Q® Water Purification Solutions