• USA Home
  • P0807 - Polyvinylidene difluoride membranes

EMAIL THIS PAGE TO A FRIEND
P0807 Supelco

Polyvinylidene difluoride membranes

L × W 20 cm × 20 cm

Synonym: PVDF

Purchase

Properties

mfr. no.   S&S 10485288
L × W   20 cm × 20 cm
pore size   0.45 μm
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?