EMAIL THIS PAGE TO A FRIEND
P8583 Sigma-Aldrich

Palatinitol

≥98%

Synonym: Palatinit

  • CAS Number 64519-82-0

  • Empirical Formula (Hill Notation) C12H24O11

  • Molecular Weight 344.31

  •  MDL number MFCD00190708

Purchase

Properties

assay   ≥98%

Description

Other Notes

A 1:1 mixture of 6-O-α-D-glucopyranosyl-D-glucitol (isomaltitol) and 6-O-α-D-glucopyranosyl-D-mannitol,

Preparation Note

Produced by reduction of palatinose.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?