• USA Home
  • PZ0009 - Tolterodine L-tartrate

EMAIL THIS PAGE TO A FRIEND
PZ0009 Sigma

Tolterodine L-tartrate

≥98% (HPLC)

Synonym: 2-[(1R)-3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-methyl-phenol tartrate, Detrusitol, PNU-200583E

Purchase

Properties

Related Categories Antagonists, Approved Therapeutics/Drug Candidates, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology,
InChI Key   TWHNMSJGYKMTRB-KXYUELECSA-N
assay   ≥98% (HPLC)
form   powder
color   white to off-white
solubility   DMSO: ≥20 mg/mL
originator   Pfizer
storage temp.   room temp

Description

Biochem/physiol Actions

Tolterodine L-tartrate is a bladder-specific muscarinic antagonist.

Other Notes

This compound was developed by Pfizer for Neuroscience research. To learn more about Sigma′s partnership with Pfizer and view other authentic, high-quality Pfizer compounds, visit sigma.com/bsm-pfizer.

To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

5, 25 mg in glass bottle

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®Pfizer), a collection of 90 Pfizer-developed drugs and research tools. Click here to learn more.

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Discover your Target

Making Headway
Protocols & Articles

Articles

Acetylcholine Receptors (Muscarinic)

Muscarinic acetylcholine receptors are members of the superfamily of G protein-coupled receptors (GPCRs). They are relatively abundant and mediate many of the diverse actions of acetylcholine in the ...
Keywords: Alzheimer Disease, Apoptosis, Atomic absorption spectroscopy, Gene expression, Hormones, Molecular biology, Neurotransmitters, Parkinson Disease, Polymorphisms, Transduction

Discover Bioactive Small Molecules for Neuroscience

The nervous system, comprised of the brain and spinal cord (central nervous system, or CNS) and network of nerves that connects the CNS to the rest of the body (peripheral nervous system, or PNS) is ...
Keywords: Bioactive small molecules, Central Nervous System, Diseases, Metabolism, Neurodegenerative Diseases, Neuroscience, Neurotransmission, Neurotransmitters

Pfizer Compounds for Neuroscience

From our library of Articles, Sigma-Aldrich presents Pfizer Compounds for Neuroscience

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?