• USA Home
  • PZ0011 - Eletriptan hydrobromide

EMAIL THIS PAGE TO A FRIEND
PZ0011 Sigma

Eletriptan hydrobromide

≥98% (HPLC)

  • CAS Number 177834-92-3

  • Empirical Formula (Hill Notation) C22H26N2O2S · HBr

  • Molecular Weight 463.43

  •  MDL number MFCD08141806

  •  PubChem Substance ID 329823680

Purchase

Description

Biochem/physiol Actions

Eletriptan hydrobromide is a serotonin 5-HT1B/1D receptor agonist; second generation anti-migraine drug.

Other Notes

This compound was developed by Pfizer for Neuroscience research. To learn more about Sigma′s partnership with Pfizer and view other authentic, high-quality Pfizer compounds, visit sigma.com/bsm-pfizer.

To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

5, 25 mg in glass bottle

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®Pfizer), a collection of 90 Pfizer-developed drugs and research tools. Click here to learn more.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Discover your Target

Making Headway
Protocols & Articles

Articles

Pfizer Compounds for Neuroscience

From our library of Articles, Sigma-Aldrich presents Pfizer Compounds for Neuroscience

Serotonin Receptors

Serotonin (5-hydroxytryptamine, 5-HT) is widely distributed throughout the mammalian body, being synthesized from L-tryptophan in enterochromaffin cells of the gastro-intestinal tract as well as in s...
Keywords: Active transport, Atomic absorption spectroscopy, Cancer, Clinical, Gene expression, Ligands, Neurotransmitters, Obesity, Polymorphisms, Schizophrenia, Transduction

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?