• USA Home
  • S0196 - Stemline® Keratinocyte Medium II

EMAIL THIS PAGE TO A FRIEND
S0196 Sigma-Aldrich

Stemline® Keratinocyte Medium II

Serum-free, Without L-glutamine, calcium-free, with HEPES, Sodium Bicarbonate, and phenol red, liquid, sterile-filtered

Synonym: Extended Life Span keratinocyte medium

Purchase

Properties

Related Categories 3D Stem Cell Culture using 3D Biotek 3D Insert Scaffolds, Cell Biology, Cell Culture, Cell Culture Supplies, Culture,
Quality Level   200
description   Requires supplementation: S9945, G8541, G7513, G6392, C7902
sterility   sterile-filtered
form   liquid
quality   Serum-free
  Without L-glutamine, calcium-free, with HEPES, Sodium Bicarbonate, and phenol red
storage temp.   2-8°C

Description

General description

Developed to promote the optimal expansion of human epidermal keratinocytes from adult and neonatal sources, Stemline Keratinocyte Medium II performs most effectively when supplemented with either Stemline Keratinocyte Growth Supplement (S9945) or Keratinocyte Medium Supplement, BPE-free (K3136).

Stemline Keratinocyte Medium II is free of serum and all other animal components; this exclusion increases performance consistency and elimates safety risks associated with potential adventitious agents.

Produced in a GMP state-of-the-art facility, Stemline Keratinocyte Stem Cell Expansion Medium is clearly an excellent choice for your Keratinocyte applications.

Application

Stemline® Keratinocyte Medium II has been used to culture explants for keratocytes.

Legal Information

Stemgent is a registered trademark of Stemgent, Inc.

Stemline is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Neural Stem Cell Culture

Neural Stem Cell Lines Neural Stem Cell Media and Supplements Neural Stem Cell Markers Stem Cell Culture Protocols
Keywords: Cell biology, Cell culture, Central Nervous System, Cryopreservation, Diseases, Dopamine agents, Environmental, Gene expression, Growth factors, Neuroscience, Phase transitions, Stem cell biology, Transcription

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?